CAS RN 13478-93-8
IUPAC Name:
bis(N-(3-nitrosobut-2-en-2-yl)hydroxylamine) nickel
Synonyms:
bis(N-(3-nitrosobut-2-en-2-yl)hydroxylamine) nickel
Molecular Weight (g/mol):
290.93
Molecular Formula:
C8H16N4NiO4
InChi Key:
CNOULSXJEKYGTK-UHFFFAOYSA-N
SMILES:
[Ni].CC(NO)=C(C)N=O.CC(NO)=C(C)N=O
1
–
1
of
1
results
1
Nickel dimethylglyoxime, 99%, Thermo Scientific™
CAS: 13478-93-8 Molecular Formula: C8H14N4NiO4 Molecular Weight (g/mol): 288.92 MDL Number: MFCD00136028 InChI Key: UNMGLSGVXHBBPH-USBCBBGOSA-L Synonym: nickel dimethylglyoxime,,nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine PubChem CID: 15639912 SMILES: [Ni++].C\C(=N\O)\C(\C)=N/[O-].C\C(=N\O)\C(\C)=N/[O-]