accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1347814-95-2

IUPAC Name: {5-chlorofuro[3,2-b]pyridin-2-yl}methanol
Synonyms: {5-chlorofuro[3,2-b]pyridin-2-yl}methanol
Molecular Weight (g/mol): 183.59
Molecular Formula: C8H6ClNO2
InChi Key: KEGRTUBZRMQESQ-UHFFFAOYSA-N
SMILES: OCC1=CC2=NC(Cl)=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results