accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONNNNN

CAS RN 1347815-06-8

IUPAC Name: tert-butyl 4,5-dicyano-2-(pyridin-3-yl)-1H-imidazole-1-carboxylate
Synonyms: tert-butyl 4,5-dicyano-2-(pyridin-3-yl)imidazole-1-carboxylate
Molecular Weight (g/mol): 295.3
Molecular Formula: C15H13N5O2
InChi Key: VHYAIVBSALAEJF-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C(=NC(C#N)=C1C#N)C1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results