accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 134855-89-3

H3COHNH2HBr

CAS RN 134855-89-3

IUPAC Name: 4-(1-aminoethyl)phenol hydrobromide
Synonyms: 1-(p-hydroxyphenyl)ethylamine hydrobromide
Molecular Weight (g/mol): 218.09
Molecular Formula: C8H12BrNO
InChi Key: PZBBMKOZPQAHRA-UHFFFAOYNA-N
SMILES: Br.CC(N)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results