CAS RN 134862-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
meta-Chlorambucil, TRC
CAS: 134862-11-6 Molecular Formula: C14H19Cl2NO2 Molecular Weight (g/mol): 304.21 Synonym: 3-[Bis(2-chloroethyl)amino]benzenebutanoic Acid,Chlorambucil Impurity IUPAC Name: 4-[3-[bis(2-chloroethyl)amino]phenyl]butanoic acid SMILES: OC(=O)CCCc1cccc(c1)N(CCCl)CCCl