CAS RN 13494-80-9
Tellurium shot, 2-5mm (0.08-0.20in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium, 99.8%, (trace metal basis), powder, -200 mesh
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium, 99+%, (trace metal basis), powder
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium powder, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium lump, 99.999+% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium pieces, 99.9999% (metals basis), zone refined, Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium powder, -18+60 mesh, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium broken ingot, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium broken ingot, Puratronic™, 99.9999+% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium lump, typically 12mm (0.47 in.), 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
Tellurium powder, -30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 13494-80-9 Molecular Formula: Te Molecular Weight (g/mol): 127.60 MDL Number: MFCD00134062 InChI Key: PORWMNRCUJJQNO-UHFFFAOYSA-N Synonym: tellurium, elemental,telloy,tellur,aurum paradoxum,element,tellur polish,metallum problematum,tellurium, metallic,unii-nqa0o090zj,and compounds PubChem CID: 6327182 ChEBI: CHEBI:30452 IUPAC Name: tellurium SMILES: [Te]
- 1
- 2