accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 135-02-4

IUPAC Name: 2-methoxybenzaldehyde
Synonyms: O-anisaldehyde
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: PKZJLOCLABXVMC-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)