accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 135-15-9

IUPAC Name: 1,4-dibutoxy-2-nitrobenzene
Synonyms: 1,4-dibutoxy-2-nitrobenzene
Molecular Weight (g/mol): 267.33
Molecular Formula: C14H21NO4
InChi Key: UVLGMZCLSQBSPZ-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(OCCCC)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results