CAS RN 1350352-70-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Descyano-3-((hydroxyimino)methyl) Febuxostat, TRC
CAS: 1350352-70-3 Molecular Formula: C16H18N2O4S Molecular Weight (g/mol): 334.39 Synonym: 2-[3-[(Hydroxyimino)methyl]-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic acid SMILES: CC(C)COc1ccc(cc1\C=N/O)c2nc(C)c(s2)C(=O)O