CAS RN 1350514-68-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Grazoprevir, TRC
CAS: 1350514-68-9 Synonym: (1R,2S)-N-[[[(1R,2R)-2-[5-(3-Hydroxy-6-methoxy-2-quinoxalinyl)pentyl]cyclopropyl]oxy]carbonyl]-3-methyl-L-valyl-(4R)-4-hydroxy-L-prolyl-1-amino-N-(cyclopropylsulfonyl)-2-ethenylcyclopropanecarboxamide Cyclic (1→2)-Ether∣Grazoprevir