accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1350626-27-5

BOOH3CCH3H3CH3CNN

CAS RN 1350626-27-5

IUPAC Name: 1-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Synonyms: 1-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 334.23
Molecular Formula: C20H23BN2O2
InChi Key: TXYJAQWDYBJALP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(CC2=CC=CC=C2)C2=NC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results