accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13510-42-4

RbOClOOO

CAS RN 13510-42-4

IUPAC Name: rubidium(1+) perchlorate
Synonyms: rubidium perchlorate
Molecular Weight (g/mol): 184.91
Molecular Formula: ClO4Rb
InChi Key: NQYGGOOZLKJKPS-UHFFFAOYSA-M
SMILES: [Rb+].[O-][Cl](=O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results