accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)OOHNHOO

CAS RN 135112-28-6

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 339.39
Molecular Formula: C20H21NO4
InChi Key: JBIJSEUVWWLFGV-UHFFFAOYNA-N
SMILES: CCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results