CAS RN 1351692-92-6
CAS RN 1351692-92-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Febuxostat Acyl-Beta-D-glucuronide, TRC
CAS: 1351692-92-6 Molecular Formula: C22 H24 N2 O9 S Molecular Weight (g/mol): 492.5 Synonym: 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylate β-D-Glucopyranuronic Acid,2-(3-Cyano-4-isobutyloxyphenyl)-4-methyl- 5-thiazolecarboxylate β-D-Glucopyranuronic Acid IUPAC Name: (2S,3S,4S,5R,6S)-6-[2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carbonyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: CC(C)COc1ccc(cc1C#N)c2nc(C)c(s2)C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O