accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OBrBrBrSm

CAS RN 13517-12-9

IUPAC Name: samarium(3+) hexahydrate tribromide
Synonyms: samarium(3+) hexahydrate tribromide
Molecular Weight (g/mol): 498.16
Molecular Formula: Br3H12O6Sm
InChi Key: LXJDCZVUWJNXRL-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results