accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13531-48-1

IUPAC Name: methyl 3-cyanobenzoate
Synonyms: methyl 3-cyanobenzoate
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: XPBHWSMZTSSEJE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results