accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClTm

CAS RN 13537-18-3

IUPAC Name: thulium(3+) trichloride
Synonyms: thulium(III) chloride
Molecular Weight (g/mol): 275.28
Molecular Formula: Cl3Tm
InChi Key: ILOTUXNTERMOJL-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Tm+3]
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results