accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1354359-53-7

CH3H3COOHN

CAS RN 1354359-53-7

IUPAC Name: 2-(propan-2-yl)phenyl N-[2-(naphthalen-2-yl)ethyl]carbamate
Synonyms: 2-isopropylphenyl N-[2-(naphthalen-2-yl)ethyl]carbamate
Molecular Weight (g/mol): 333.43
Molecular Formula: C22H23NO2
InChi Key: PCNJGBMWAZRVEA-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1OC(=O)NCCC1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results