accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNFFF

CAS RN 13544-43-9

IUPAC Name: 6-(trifluoromethyl)-1H-indole
Synonyms: 6-(trifluoromethyl)-1H-indole
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: BPYBYPREOVLFED-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C2C=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results