accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBr

CAS RN 135613-33-1

IUPAC Name: ethyl 3-(2-bromophenyl)propanoate
Synonyms: ethyl 3-(2-bromophenyl)propanoate
Molecular Weight (g/mol): 257.13
Molecular Formula: C11H13BrO2
InChi Key: ZKICRDRBPPOQLM-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results