accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13568-40-6

LiLiOMoOOO

CAS RN 13568-40-6

IUPAC Name: dilithium(1+) dioxomolybdenumbis(olate)
Synonyms: lithium molybdate
Molecular Weight (g/mol): 173.83
Molecular Formula: Li2MoO4
InChi Key: NMHMDUCCVHOJQI-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[O-][Mo]([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results