accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13569-80-7

FFFDy

CAS RN 13569-80-7

IUPAC Name: dysprosium(3+) trifluoride
Synonyms: dysprosium(3+) trifluoride
Molecular Weight (g/mol): 219.5
Molecular Formula: DyF3
InChi Key: FWQVINSGEXZQHB-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Dy+3]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)