CAS RN 13587-19-4
CAS RN 13587-19-4
IUPAC Name:
dicaesium(1+) tungsten tetraoxidandiide
Synonyms:
caesium tungstate
Molecular Weight (g/mol):
513.65
Molecular Formula:
Cs2O4W
InChi Key:
ZYAJNDITUUBDPX-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[Cs+].[Cs+].[W]
1
–
1
of
1
results
1
Cesium tungsten oxide, 99.9% (metals basis)
CAS: 13587-19-4 Molecular Formula: Cs2O4W MDL Number: MFCD00049438 Synonym: Cesium tungstate