accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 135906-00-2

NOOHO

CAS RN 135906-00-2

IUPAC Name: 1-cyclohexyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: 1-cyclohexyl-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol): 271.32
Molecular Formula: C16H17NO3
InChi Key: DTLUKEHVTWNAGE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN(C2CCCCC2)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results