accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClClClRe

CAS RN 13596-35-5

IUPAC Name: rhenium(5+) pentachloride
Synonyms: rhenium pentachloride
Molecular Weight (g/mol): 363.46
Molecular Formula: Cl5Re
InChi Key: XZQYTGKSBZGQMO-UHFFFAOYSA-I
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Re+5]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results