accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13598-36-2

IUPAC Name: dihydroxy(oxo)-λ⁵-phosphanylium
Synonyms: phosphorous acid
Molecular Weight (g/mol): 80.99
Molecular Formula: H2O3P
InChi Key: XNQULTQRGBXLIA-UHFFFAOYSA-O
SMILES: O[P+](O)=O
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Grade 
  • (4)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (5)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)