accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13598-65-7

NH4OReOOO

CAS RN 13598-65-7

IUPAC Name: ammonium tetraoxorheniumuide
Synonyms: ammonium perrhenate
Molecular Weight (g/mol): 268.24
Molecular Formula: H4NO4Re
InChi Key: NLESJKWOAILDGM-UHFFFAOYSA-O
SMILES: [NH4+].O=[Re-](=O)(=O)=O
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)