accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHNN

CAS RN 136-85-6

IUPAC Name: 5-methyl-2H-1,2,3-benzotriazole
Synonyms: 5-methyl-2H-1,2,3-benzotriazole
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: LRUDIIUSNGCQKF-UHFFFAOYSA-N
SMILES: CC1=CC2=NNN=C2C=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)