accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13601-06-4

IUPAC Name: palladium(2+) tetraamine sulfate
Synonyms: palladium(2+) tetraamine sulfate
Molecular Weight (g/mol): 270.6
Molecular Formula: H12N4O4PdS
InChi Key: WSPHQZJZSJENIO-UHFFFAOYSA-L
SMILES: N.N.N.N.[Pd++].[O-]S([O-])(=O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results