accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(S)NOO

CAS RN 136030-33-6

IUPAC Name: (3S)-2-{[(9H-fluoren-9-yl)methoxy]carbonyl}-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms: (3S)-2-[(9H-fluoren-9-ylmethoxy)carbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylate
Molecular Weight (g/mol): 398.44
Molecular Formula: C25H20NO4
InChi Key: LIRBCUNCXDZOOU-QHCPKHFHSA-M
SMILES: [O-]C(=O)[C@@H]1CC2=CC=CC=C2CN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results