accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH

CAS RN 13605-19-1

IUPAC Name: 2-(3-methylphenoxy)ethan-1-ol
Synonyms: ETHYLENEGLYCOLMONO3methylphenylether
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: FDBXUXVQIOQYIX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(OCCO)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results