accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFF

CAS RN 136239-66-2

IUPAC Name: 1-butoxy-2,3-difluorobenzene
Synonyms: 1-butoxy-2,3-difluorobenzene
Molecular Weight (g/mol): 186.2
Molecular Formula: C10H12F2O
InChi Key: HYDIPEHDSUNHPA-UHFFFAOYSA-N
SMILES: CCCCOC1=C(F)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results