accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 136459-72-8

CH3SiH3CCH3

CAS RN 136459-72-8

IUPAC Name: trimethyl[4-(2-phenylethynyl)phenyl]silane
Synonyms: trimethyl[4-(2-phenylethynyl)phenyl]silane
Molecular Weight (g/mol): 250.42
Molecular Formula: C17H18Si
InChi Key: UZRJWRFLYPENFH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=C(C=C1)C#CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results