accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)(S)(S)(S)O(R)(R)CH3NHO(S)(S)OH

CAS RN 13647-35-3

IUPAC Name: (1S,2R,6R,8S,11S,12S,15S,16S)-5,15-dihydroxy-2,16-dimethyl-7-oxapentacyclo[9.7.0.0²,⁸.0⁶,⁸.0¹²,¹⁶]octadec-4-ene-4-carbonitrile
Synonyms: trilostane modrenal modrastane desopan trilostano trilostane (product) trilostanum trilostane preparation trilostane (substance) wanskerne brand of trilostane
Molecular Weight (g/mol): 329.44
Molecular Formula: C20H27NO3
InChi Key: KVJXBPDAXMEYOA-CXANFOAXSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CC[C@@]45O[C@@H]4C(O)=C(C[C@]35C)C#N)[C@@H]1CC[C@@H]2O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results