accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrHNBr

CAS RN 136630-39-2

IUPAC Name: 2,7-dibromo-9H-carbazole
Synonyms: 2,7-dibromo-9H-carbazole
Molecular Weight (g/mol): 325
Molecular Formula: C12H7Br2N
InChi Key: QPTWWBLGJZWRAV-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results