accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 136663-21-3

IUPAC Name: methyl 2-methyl-1,3-benzoxazole-5-carboxylate
Synonyms: methyl 2-methyl-1,3-benzoxazole-5-carboxylate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: UFLDNRAXHGPDOD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2OC(C)=NC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results