accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 136725-55-8

IUPAC Name: 3-fluoropyrrolidine hydrochloride
Synonyms: 3-fluoropyrrolidine hydrochloride
Molecular Weight (g/mol): 125.57
Molecular Formula: C4H9ClFN
InChi Key: LENYOXXELREKGZ-UHFFFAOYNA-N
SMILES: Cl.FC1CCNC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)