accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13677-79-7

IUPAC Name: 3,4,5-trihydroxybenzaldehyde
Synonyms: 3,4,5-trihydroxybenzaldehyde
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: RGZHEOWNTDJLAQ-UHFFFAOYSA-N
SMILES: OC1=CC(C=O)=CC(O)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results