accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 136779-28-7

IUPAC Name: 1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane
Synonyms: 1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane
Molecular Weight (g/mol): 362.48
Molecular Formula: C22H36P2
InChi Key: GVVCHDNSTMEUCS-UHFFFAOYNA-N
SMILES: CCC1CCC(CC)P1C1=CC=CC=C1P1C(CC)CCC1CC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results