accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSOCH3

CAS RN 13678-59-6

IUPAC Name: 2-methyl-5-(methylsulfanyl)furan
Synonyms: 2-methyl-5-(methylsulfanyl)furan
Molecular Weight (g/mol): 128.19
Molecular Formula: C6H8OS
InChi Key: RESBOJMQOGJOMW-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results