accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2H3COH2C

CAS RN 13679-86-2

IUPAC Name: 2-ethenyl-2-methyl-5-(prop-1-en-2-yl)oxolane
Synonyms: 2-ethenyl-2-methyl-5-(prop-1-en-2-yl)oxolane
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: XIGFNCYVSHOLIF-UHFFFAOYNA-N
SMILES: CC(=C)C1CCC(C)(O1)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)