accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OBOCH3N

CAS RN 136805-54-4

IUPAC Name: dimethyl (pyridin-2-yl)boronate
Synonyms: dimethyl pyridin-2-ylboronate
Molecular Weight (g/mol): 150.97
Molecular Formula: C7H10BNO2
InChi Key: ITSIDKYMJQVPQQ-UHFFFAOYSA-N
SMILES: COB(OC)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results