accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BH2CFFFK

CAS RN 13682-77-4

IUPAC Name: potassium ethenyltrifluoroboranuide
Synonyms: potassium ethenyltrifluoroboranuide
Molecular Weight (g/mol): 133.95
Molecular Formula: C2H3BF3K
InChi Key: ZCUMGICZWDOJEM-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C=C
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results