accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiCH3H3COOH2C

CAS RN 13688-55-6

IUPAC Name: trimethylsilyl prop-2-enoate
Synonyms: trimethylsilyl prop-2-enoate
Molecular Weight (g/mol): 144.25
Molecular Formula: C6H12O2Si
InChi Key: OTYBJBJYBGWBHB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results