accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13688-90-9

ClSiClClCl

CAS RN 13688-90-9

IUPAC Name: trichloro[4-(chloromethyl)phenyl]silane
Synonyms: trichloro[4-(chloromethyl)phenyl]silane
Molecular Weight (g/mol): 260.01
Molecular Formula: C7H6Cl4Si
InChi Key: YBYBQFPZMKPGPJ-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results