accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(R)OOONHOHCH3H3CHO(R)OOONHOHH3CCH3HOCa

CAS RN 137-08-6

IUPAC Name: calcium bis(3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate)
Synonyms: calcium pantothenate
Molecular Weight (g/mol): 476.54
Molecular Formula: C18H32CaN2O10
InChi Key: FAPWYRCQGJNNSJ-UHFFFAOYNA-L
SMILES: [Ca++].CC(C)(CO)C(O)C(=O)NCCC([O-])=O.CC(C)(CO)C(O)C(=O)NCCC([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (8)
  • (2)
Quantity 
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (6)
  • (8)
  • (2)
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)