accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Br

CAS RN 137-43-9

IUPAC Name: bromocyclopentane
Synonyms: bromocyclopentane
Molecular Weight (g/mol): 149.03
Molecular Formula: C5H9Br
InChi Key: BRTFVKHPEHKBQF-UHFFFAOYSA-N
SMILES: BrC1CCCC1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (5)