accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNSHNCH3

CAS RN 137-97-3

IUPAC Name: 1,3-bis(2-methylphenyl)thiourea
Synonyms: 1,3-bis(2-methylphenyl)thiourea
Molecular Weight (g/mol): 256.37
Molecular Formula: C15H16N2S
InChi Key: KWPNNZKRAQDVPZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(=S)NC1=CC=CC=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results