accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 137052-08-5

IUPAC Name: 1-(oxan-4-yl)ethan-1-one
Synonyms: 1-(oxan-4-yl)ethanone
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: VNMXIOWPBADSIC-UHFFFAOYSA-N
SMILES: CC(=O)C1CCOCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results