accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFTb

CAS RN 13708-63-9

IUPAC Name: terbium(3+) trifluoride
Synonyms: terbium(3+) trifluoride
Molecular Weight (g/mol): 215.92
Molecular Formula: F3Tb
InChi Key: LKNRQYTYDPPUOX-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Tb+3]
Grade 
  • (5)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)